Assay Details
| Gene Symbol: Sox17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289464.1 | NP_001276393.1 | 1 - 2 | 1068 | 121 | |
| NM_001289467.1 | NP_001276396.1 | 1 - 2 | 1068 | 121 | ||
| GenBank mRNA | D49473.1 | - | 1 - 2 | 477 | 121 | |
| D49474.1 | - | 1 - 2 | 77 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D49473.1, D49474.1, NM_001289464.1, NM_001289467.1 transcripts.
Set Membership: |
|

Sox17
Genomic Map