Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 10 - 11 | 1455 | 92 | |
| NM_001290721.1 | NP_001277650.1 | 9 - 10 | 1262 | 92 | ||
| NM_207228.3 | NP_997111.1 | 10 - 11 | 1322 | 92 | ||
| GenBank mRNA | AF530050.3 | - | 9 - 10 | 1267 | 92 | |
| AK030254.1 | - | 10 - 11 | 1455 | 92 | ||
| AY618264.1 | - | 9 - 10 | 1267 | 92 | ||
| BC066782.1 | - | 10 - 11 | 1319 | 6705127 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsga10
Genomic Map