Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 14 - 15 | 1935 | 88 | |
| NM_001290721.1 | NP_001277650.1 | 13 - 14 | 1742 | 88 | ||
| NM_207228.3 | NP_997111.1 | 14 - 15 | 1802 | 88 | ||
| NR_110976.1 | 12 - 13 | 1587 | 88 | |||
| GenBank mRNA | AF530050.3 | - | 13 - 14 | 1747 | 88 | |
| AK030254.1 | - | 14 - 15 | 1935 | 88 | ||
| AY618264.1 | - | 13 - 14 | 1747 | 88 | ||
| BC053407.1 | - | 12 - 13 | 1570 | 88 | ||
| BC066782.1 | - | 14 - 15 | 1799 | 6705127 | 88 | |
| CK789086.1 | - | 3 - 4 | 311 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tsga10
Genomic Map