Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 2 - 3 | 317 | 132 | |
| NM_001290721.1 | NP_001277650.1 | 1 - 2 | 124 | 132 | ||
| NM_207228.3 | NP_997111.1 | 2 - 3 | 184 | 132 | ||
| NR_110976.1 | 1 - 2 | 124 | 132 | |||
| GenBank mRNA | AK030254.1 | - | 2 - 3 | 318 | 131 | |
| AK163138.1 | - | 2 - 3 | 175 | 132 | ||
| AK164954.1 | - | 2 - 3 | 149 | 132 | ||
| AK170053.1 | - | 2 - 3 | 207 | 132 | ||
| BC053407.1 | - | 1 - 2 | 107 | 132 | ||
| BC066782.1 | - | 2 - 3 | 181 | 6705127 | 132 | |
| CJ046680.1 | - | 2 - 3 | 180 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163138.1, BC066782.1, NM_001290720.1, NM_001290721.1, NM_207228.3 transcripts.
Set Membership: |
|

Tsga10
Genomic Map