Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 3 - 4 | 530 | 139 | |
| NM_001290721.1 | NP_001277650.1 | 2 - 3 | 337 | 139 | ||
| NM_207228.3 | NP_997111.1 | 3 - 4 | 397 | 139 | ||
| NR_110976.1 | 2 - 3 | 337 | 139 | |||
| GenBank mRNA | AF530050.3 | - | 2 - 3 | 339 | 139 | |
| AK030254.1 | - | 3 - 4 | 530 | 139 | ||
| AK164954.1 | - | 3 - 4 | 362 | 139 | ||
| AK170053.1 | - | 3 - 4 | 420 | 139 | ||
| AY618264.1 | - | 2 - 3 | 339 | 139 | ||
| BC053407.1 | - | 2 - 3 | 320 | 139 | ||
| BC066782.1 | - | 3 - 4 | 394 | 6705127 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF530050.3, AY618264.1, BC066782.1, NM_001290720.1, NM_001290721.1, NM_207228.3 transcripts.
Set Membership: |
|

Tsga10
Genomic Map