Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 6 - 7 | 932 | 79 | |
| NM_001290721.1 | NP_001277650.1 | 5 - 6 | 739 | 79 | ||
| NM_207228.3 | NP_997111.1 | 6 - 7 | 799 | 79 | ||
| NR_110976.1 | 5 - 6 | 739 | 79 | |||
| GenBank mRNA | AF530050.3 | - | 5 - 6 | 744 | 79 | |
| AK030254.1 | - | 6 - 7 | 932 | 79 | ||
| AK163138.1 | - | 6 - 7 | 746 | 79 | ||
| AK170053.1 | - | 6 - 7 | 822 | 79 | ||
| AY618264.1 | - | 5 - 6 | 744 | 79 | ||
| BC053407.1 | - | 5 - 6 | 722 | 79 | ||
| BC066782.1 | - | 6 - 7 | 796 | 6705127 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsga10
Genomic Map