Assay Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290720.1 | NP_001277649.1 | 8 - 9 | 1178 | 75 | |
| NM_001290721.1 | NP_001277650.1 | 7 - 8 | 985 | 75 | ||
| NM_207228.3 | NP_997111.1 | 8 - 9 | 1045 | 75 | ||
| NR_110976.1 | 7 - 8 | 985 | 75 | |||
| GenBank mRNA | AF530050.3 | - | 7 - 8 | 990 | 75 | |
| AK030254.1 | - | 8 - 9 | 1178 | 75 | ||
| AY618264.1 | - | 7 - 8 | 990 | 75 | ||
| BC053407.1 | - | 7 - 8 | 968 | 75 | ||
| BC066782.1 | - | 8 - 9 | 1042 | 6705127 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsga10
Genomic Map