Assay Details
| Gene Symbol: Mitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026913.2 | NP_081189.1 | 5 - 6 | 697 | 91 | |
| GenBank mRNA | AK171903.1 | - | 5 - 6 | 698 | 91 | |
| BC020137.1 | - | 693 | 3709157 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156459 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122110 | AK171903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122110 | BC020137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122110 | NM_026913.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122110 | AK005342.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120821 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121115 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122110 | AK171903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122110 | BC020137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122110 | NM_026913.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122110 | AK005342.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04067209 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00047217 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121115 | - | - |
| Whole Mouse Genome G4122A | A_51_P303332 | DV649848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303332 | NM_001037918.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303332 | AK048005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303332 | BC019519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303332 | BC020137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P203833 | AK005342.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P203837 | AK171903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P203837 | BC020137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P203837 | NM_026913.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P203837 | AK005342.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00578507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00578511 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00578504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00243507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00139206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00397557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01103105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00669458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00139207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01056599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00660351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01056596 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10354058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354059 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426133_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439261_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354054 | - | - |
| Mouse Genome 430 2.0 Array | 1439261_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439261_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426133_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426133_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69028_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69028_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100017806.mm.1 | NM_026913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017806.mm.1 | NM_026913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017806.mm.1 | TC0100002210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100017807.mm.1 | NM_026913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017807.mm.1 | NM_026913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017807.mm.1 | TC0100002210.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100017804.mm.1 | NM_026913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017804.mm.1 | NM_026913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017804.mm.1 | TC0100002210.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426133_PM_a_at | - | - |
| Mouse Expression Set 430 | 1439261_x_at | - | - |
| Mouse Expression Set 430 | 1439261_PM_x_at | - | - |
| Mouse Expression Set 430 | 1426133_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439261_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426133_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426133_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439261_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426133_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426133_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439261_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439261_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 385915 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217646 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217646 | - | - |

Mitd1
Genomic Map