Assay Details
| Gene Symbol: Lpar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175271.4 | NP_780480.2 | 2 - 3 | 471 | 76 | |
| GenBank mRNA | AK045289.1 | - | 2 - 3 | 441 | 76 | |
| AK051709.1 | - | 2 - 3 | 442 | 76 | ||
| BC052178.1 | - | 2 - 3 | 462 | 76 | ||
| BC069178.1 | - | 2 - 3 | 383 | 76 | ||
| BC138893.1 | - | 2 - 3 | 272 | 8861915 | 76 | |
| BC138894.1 | - | 2 - 3 | 272 | 8861916 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045289.1, AK051709.1, BC138893.1, BC138894.1, NM_175271.4 transcripts.
Set Membership: |
|

Lpar4
Genomic Map