Assay Details
| Gene Symbol: 4932413F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040764.1 | 2 - 3 | 2478 | 84 | ||
| GenBank mRNA | AK077024.1 | - | 2 - 3 | 2480 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 16252 | Gm16252 | OTTMUSG00000032743 | -59.0kb | Sense | Chr. 9 - 103472049 - 103503357 on NCBI Build GRCm38 |
| Downstream | predicted gene 29154 | Gm29154 | 353.5kb | Anti-sense | Chr. 9 - 103919392 - 103960323 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4932413F04Rik
Genomic Map