Assay Details
| Gene Symbol: Cebpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024806.2 | NP_001019977.1 | 12 - 13 | 2837 | 122 | |
| GenBank mRNA | AK136173.1 | - | 12 - 13 | 2818 | 122 | |
| AK145374.1 | - | 12 - 13 | 2819 | 122 | ||
| AK155990.1 | - | 12 - 13 | 2820 | 122 | ||
| AK158771.1 | - | 12 - 13 | 2814 | 122 | ||
| AK159081.1 | - | 12 - 13 | 2834 | 122 | ||
| BB704756.1 | - | 4 - 5 | 427 | 122 | ||
More Information
Set Membership: |
|

Cebpz
Genomic Map