Assay Details
| Gene Symbol: Cebpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024806.2 | NP_001019977.1 | 7 - 8 | 2352 | 86 | |
| GenBank mRNA | AK136173.1 | - | 7 - 8 | 2333 | 86 | |
| AK145374.1 | - | 7 - 8 | 2334 | 86 | ||
| AK155990.1 | - | 7 - 8 | 2335 | 86 | ||
| AK158771.1 | - | 7 - 8 | 2329 | 86 | ||
| AK159081.1 | - | 7 - 8 | 2349 | 86 | ||
| U19891.1 | - | 7 - 8 | 2323 | 86 | ||
| U19892.1 | - | 7 - 8 | 2037 | 86 | ||
More Information
Set Membership: |
|

Cebpz
Genomic Map