Assay Details
| Gene Symbol: Cebpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024806.2 | NP_001019977.1 | 9 - 10 | 2490 | 99 | |
| GenBank mRNA | AK136173.1 | - | 9 - 10 | 2471 | 99 | |
| AK145374.1 | - | 9 - 10 | 2472 | 99 | ||
| AK155990.1 | - | 9 - 10 | 2473 | 99 | ||
| AK158771.1 | - | 9 - 10 | 2467 | 99 | ||
| AK159081.1 | - | 9 - 10 | 2487 | 99 | ||
| U19891.1 | - | 9 - 10 | 2461 | 99 | ||
| U19892.1 | - | 9 - 10 | 2175 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cebpz
Genomic Map