Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 26 - 27 | 4256 | 66 | |
| GenBank mRNA | AB299229.1 | - | 26 - 27 | 4183 | 66 | |
| AK046668.1 | - | 5 - 6 | 745 | 66 | ||
| BC047418.1 | - | 16 - 17 | 2369 | 66 | ||
| BC094568.1 | - | 8 - 9 | 1540 | 66 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map