Assay Details
| Gene Symbol: Il2rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143669.1 | - | 1 - 2 | 130 | 84 | |
| AK153873.1 | - | 1 - 2 | 144 | 84 | ||
| BC014720.1 | - | 1 - 2 | 124 | 4924951 | 84 | |
| D13565.1 | - | 1 - 2 | 122 | 84 | ||
| D13821.1 | - | 1 - 2 | 117 | 84 | ||
| KC815469.1 | - | 1 - 2 | 120 | 84 | ||
| X75337.1 | - | 1 - 2 | 147 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il2rg
Genomic Map