Assay Details
| Gene Symbol: Il2rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143669.1 | - | 6 - 7 | 876 | 134 | |
| AK153873.1 | - | 6 - 7 | 891 | 134 | ||
| BC014720.1 | - | 6 - 7 | 870 | 4924951 | 134 | |
| D13565.1 | - | 6 - 7 | 868 | 134 | ||
| D13821.1 | - | 6 - 7 | 863 | 134 | ||
| KC815469.1 | - | 6 - 7 | 762 | 134 | ||
| L20048.1 | - | 6 - 7 | 880 | 134 | ||
| X75337.1 | - | 6 - 7 | 893 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il2rg
Genomic Map