Assay Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198021.2 | NP_932138.1 | 11 - 12 | 1789 | 123 | |
| GenBank mRNA | AK139747.1 | - | 3 - 4 | 378 | 123 | |
| AK139761.1 | - | 11 - 12 | 1776 | 123 | ||
| BC042443.1 | - | 11 - 12 | 1789 | 4037878 | 123 | |
| BC042770.1 | - | 6 - 7 | 688 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scyl2
Genomic Map