Assay Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198021.2 | NP_932138.1 | 12 - 13 | 1914 | 85 | |
| GenBank mRNA | AK139747.1 | - | 4 - 5 | 503 | 85 | |
| AK139761.1 | - | 12 - 13 | 1901 | 85 | ||
| BC042443.1 | - | 12 - 13 | 1914 | 4037878 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scyl2
Genomic Map