Assay Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198021.2 | NP_932138.1 | 14 - 15 | 2134 | 91 | |
| GenBank mRNA | AK139747.1 | - | 6 - 7 | 723 | 91 | |
| AK139761.1 | - | 14 - 15 | 2121 | 91 | ||
| BC042443.1 | - | 14 - 15 | 2134 | 4037878 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scyl2
Genomic Map