Assay Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198021.2 | NP_932138.1 | 15 - 16 | 2204 | 69 | |
| GenBank mRNA | BC042443.1 | - | 15 - 16 | 2204 | 4037878 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Scyl2
Genomic Map