| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000020279.mm.1 |
NM_198021.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020279.mm.1 |
NM_198021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020279.mm.1 |
TC1000002659.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020278.mm.1 |
NM_198021.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020278.mm.1 |
NM_198021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020278.mm.1 |
TC1000002659.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020277.mm.1 |
NM_198021.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020277.mm.1 |
NM_198021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020277.mm.1 |
TC1000002659.mm.1 |
MTA TC |