Assay Details
Gene Symbol: Gm3942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036450.1 | - | 1 - 2 | 138 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700010J16 gene | 1700010J16Rik | -52.2kb | Sense | Chr. 10 - 112726926 - 112785599 on NCBI Build GRCm38 | |
Downstream | ataxin 7-like 3B | Atxn7l3b |
4921506J03Rik, 6230409E21Rik, AI315132, ENSMUSG00000074747 |
84.8kb | Anti-sense | Chr. 10 - 112925428 - 112929026 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
