Assay Details
| Gene Symbol: Gm4710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033456.1 | 1 - 2 | 304 | 67 | ||
| GenBank mRNA | AK082440.1 | - | 1 - 2 | 305 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 98, member A |
Fam98a |
2810405J04Rik, AL024114 |
-309.1kb | Anti-sense | Chr. 17 - 75537086 - 75551946 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32282 |
Gm32282 | 2.3mb | Anti-sense | Chr. 17 - 78199250 - 78200084 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4710
Genomic Map