Assay Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289612.1 | NP_001276541.1 | 3 - 4 | 731 | 100 | |
| NM_001289765.1 | NP_001276694.1 | 10 - 11 | 1678 | 100 | ||
| NM_001289766.1 | NP_001276695.1 | 10 - 11 | 1643 | 100 | ||
| NM_009539.3 | NP_033565.2 | 10 - 11 | 1635 | 100 | ||
| GenBank mRNA | AB083210.1 | - | 3 - 4 | 734 | 100 | |
| AB084383.1 | - | 3 - 4 | 573 | 100 | ||
| AK039883.1 | - | 10 - 11 | 1572 | 100 | ||
| AK089056.1 | - | 10 - 11 | 1607 | 100 | ||
| BC029727.1 | - | 10 - 11 | 1593 | 4925739 | 100 | |
| U04379.1 | - | 10 - 11 | 1589 | 100 | ||
| U77667.1 | - | 9 - 10 | 1481 | 100 | ||
More Information
Set Membership: |
|

Zap70
Genomic Map