Assay Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009539.3 | NP_033565.2 | 1 - 2 | 145 | 95 | |
| GenBank mRNA | AK039883.1 | - | 1 - 2 | 82 | 95 | |
| AK089056.1 | - | 1 - 2 | 118 | 95 | ||
| U04379.1 | - | 1 - 2 | 99 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039883.1, NM_009539.3, U04379.1 transcripts.
Set Membership: |
|

Zap70
Genomic Map