Assay Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289765.1 | NP_001276694.1 | 4 - 5 | 894 | 59 | |
| NM_001289766.1 | NP_001276695.1 | 4 - 5 | 859 | 59 | ||
| NM_009539.3 | NP_033565.2 | 4 - 5 | 851 | 59 | ||
| GenBank mRNA | AK039883.1 | - | 4 - 5 | 788 | 59 | |
| AK089056.1 | - | 4 - 5 | 824 | 59 | ||
| BC029727.1 | - | 4 - 5 | 809 | 4925739 | 59 | |
| U04379.1 | - | 4 - 5 | 805 | 59 | ||
| U77667.1 | - | 3 - 4 | 697 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zap70
Genomic Map