Assay Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013374.1 | NP_001013392.1 | 3 - 4 | 424 | 71 | |
| GenBank mRNA | AK041656.1 | - | 1 - 2 | 2269 | 71 | |
| AK142024.1 | - | 393 | 71 | |||
| AK158015.1 | - | 3 - 4 | 443 | 71 | ||
| BC046969.1 | - | 3 - 4 | 424 | 6313679 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lman2l
Genomic Map