Assay Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013374.1 | NP_001013392.1 | 4 - 5 | 512 | 65 | |
| GenBank mRNA | AK041656.1 | - | 2 - 3 | 2357 | 65 | |
| AK142024.1 | - | 481 | 65 | |||
| AK158015.1 | - | 4 - 5 | 531 | 65 | ||
| BC046969.1 | - | 4 - 5 | 512 | 6313679 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lman2l
Genomic Map