| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100017416.mm.1 |
NM_153408.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017416.mm.1 |
NM_153408.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017416.mm.1 |
TC0100002177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100017413.mm.1 |
NM_153408.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017413.mm.1 |
NM_153408.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017413.mm.1 |
TC0100002177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100017415.mm.1 |
NM_153408.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017415.mm.1 |
NM_153408.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017415.mm.1 |
TC0100002177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100017417.mm.1 |
NM_153408.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017417.mm.1 |
NM_153408.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017417.mm.1 |
TC0100002177.mm.1 |
MTA TC |