Assay Details
| Gene Symbol: Actr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146107.2 | NP_666219.1 | 5 - 6 | 507 | 138 | |
| GenBank mRNA | AK078508.1 | - | 5 - 6 | 523 | 138 | |
| AK088839.1 | - | 5 - 6 | 540 | 138 | ||
| AK205617.1 | - | 1 - 2 | 65 | 138 | ||
| BC023020.1 | - | 5 - 6 | 507 | 5375425 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actr1b
Genomic Map