Assay Details
| Gene Symbol: Sema4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090073.1 | - | 2 - 3 | 354 | 56 | |
| AK148298.1 | - | 2 - 3 | 355 | 56 | ||
| AK173247.1 | - | 3 - 4 | 597 | 56 | ||
| S79463.1 | - | 2 - 3 | 209 | 56 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953866 | S79463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953866 | AK173247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953866 | NM_001126047.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712639 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291016 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130093 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953866 | S79463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953866 | AK173247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953866 | NM_001126047.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170651 | S79463.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170651 | AK173247.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170651 | AK090073.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170651 | AK148298.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170651 | NM_001126047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659904 | S79463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659904 | AK173247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659904 | AK148298.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P659904 | NM_001126047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170651 | S79463.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170651 | AK173247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170651 | AK090073.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170651 | AK148298.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170651 | NM_001126047.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20149081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00560860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00560875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20149043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00266044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00560865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10353910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353907 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353904 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433920_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353899 | - | - |
| Mouse Genome 430 2.0 Array | 1433920_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444773_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433920_at | - | - |
| Mouse Genome 430 2.0 Array | 1444773_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20353_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20353_at | - | - |
| Mouse Expression Set 430 | 1433920_PM_at | - | - |
| Mouse Expression Set 430 | 1433920_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433920_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433920_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1980315 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242146 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242146 | - | - |

Sema4c
Genomic Map