Assay Details
| Gene Symbol: Sema4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148298.1 | - | 5 - 6 | 667 | 56 | |
| AK173247.1 | - | 6 - 7 | 907 | 56 | ||
| S79463.1 | - | 5 - 6 | 521 | 56 | ||
More Information
Set Membership: |
|

Sema4c
Genomic Map