Assay Details
| Gene Symbol: Bfsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002896.2 | NP_001002896.1 | 4 - 5 | 997 | 84 | |
| GenBank mRNA | AK134810.1 | - | 4 - 5 | 929 | 84 | |
| BC062815.1 | - | 2 - 3 | 199 | 84 | ||
| BC068172.1 | - | 4 - 5 | 997 | 30550909 | 84 | |
| S55549.1 | - | 2 - 3 | 306 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bfsp2
Genomic Map