Assay Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173781.4 | NP_776142.1 | 2 - 3 | 215 | 91 | |
| GenBank mRNA | AB232597.1 | - | 2 - 3 | 132 | 91 | |
| AK035893.1 | - | 2 - 3 | 174 | 91 | ||
| AK082732.1 | - | 2 - 3 | 338 | 91 | ||
| BC060618.1 | - | 2 - 3 | 215 | 6406134 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab6b
Genomic Map