Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 4 - 5 | 554 | 65 | |
| GenBank mRNA | AK004894.2 | - | 4 - 5 | 555 | 65 | |
| AK054410.1 | - | 4 - 5 | 561 | 65 | ||
| AK089235.1 | - | 4 - 5 | 558 | 65 | ||
| AK129009.1 | - | 4 - 5 | 466 | 65 | ||
| AK170421.1 | - | 4 - 5 | 547 | 65 | ||
| AK170644.1 | - | 4 - 5 | 553 | 65 | ||
| AK172467.1 | - | 4 - 5 | 516 | 65 | ||
| BC138843.1 | - | 4 - 5 | 471 | 8861865 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map