Assay Details
| Gene Symbol: Gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177910.3 | NP_808578.1 | 4 - 5 | 625 | 66 | |
| GenBank mRNA | AK088295.1 | - | 4 - 5 | 607 | 66 | |
| AK148125.1 | - | 625 | 66 | |||
| AK158470.1 | - | 4 - 5 | 625 | 66 | ||
| AK165816.1 | - | 4 - 5 | 627 | 66 | ||
| BC061207.1 | - | 5 - 6 | 499 | 30249330 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmppb
Genomic Map