Assay Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008243.3 | NP_032269.3 | 10 - 11 | 1279 | 115 | |
| GenBank mRNA | AK040934.1 | - | 10 - 11 | 1263 | 115 | |
| AK134222.1 | - | 10 - 11 | 1262 | 115 | ||
| AK144824.1 | - | 10 - 11 | 1242 | 115 | ||
| BC010551.1 | - | 10 - 11 | 1264 | 4195560 | 115 | |
| BC051393.1 | - | 10 - 11 | 1248 | 5055055 | 115 | |
| M74181.1 | - | 10 - 11 | 1236 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mst1
Genomic Map