Assay Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008243.3 | NP_032269.3 | 15 - 16 | 1784 | 60 | |
| GenBank mRNA | AK040934.1 | - | 15 - 16 | 1768 | 60 | |
| AK144824.1 | - | 15 - 16 | 1747 | 60 | ||
| BC010551.1 | - | 15 - 16 | 1769 | 4195560 | 60 | |
| BC051393.1 | - | 15 - 16 | 1753 | 5055055 | 60 | |
| M74181.1 | - | 15 - 16 | 1741 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mst1
Genomic Map