Assay Details
| Gene Symbol: Nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018879.2 | NP_061367.1 | 3 - 4 | 507 | 60 | |
| GenBank mRNA | AF131207.1 | - | 3 - 4 | 506 | 60 | |
| AK010331.1 | - | 3 - 4 | 425 | 60 | ||
| AK013299.1 | - | 3 - 4 | 469 | 60 | ||
| AK013691.1 | - | 3 - 4 | 460 | 60 | ||
| AK088925.1 | - | 3 - 4 | 472 | 60 | ||
| BC026548.1 | - | 3 - 4 | 423 | 5028027 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nprl2
Genomic Map