Assay Details
| Gene Symbol: Gm9376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101609.1 | NP_001095079.1 | 1 | 63 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0400003502.rn
Set Membership: |
|

Gm9376
Genomic Map