Assay Details
| Gene Symbol: Acad11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175324.3 | NP_780533.2 | 10 - 11 | 1318 | 78 | |
| GenBank mRNA | AK077531.1 | - | 10 - 11 | 1319 | 78 | |
| AK128998.1 | - | 10 - 11 | 1297 | 78 | ||
| AK139918.1 | - | 10 - 11 | 1262 | 78 | ||
| AK145561.1 | - | 10 - 11 | 1307 | 78 | ||
| AK145834.1 | - | 10 - 11 | 1298 | 78 | ||
| BC032994.1 | - | 3 - 4 | 247 | 78 | ||
| BC045199.1 | - | 10 - 11 | 1282 | 5133197 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acad11
Genomic Map