Assay Details
| Gene Symbol: Acad11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175324.3 | NP_780533.2 | 8 - 9 | 1129 | 83 | |
| GenBank mRNA | AK077531.1 | - | 8 - 9 | 1130 | 83 | |
| AK128998.1 | - | 8 - 9 | 1108 | 83 | ||
| AK139918.1 | - | 8 - 9 | 1073 | 83 | ||
| AK145561.1 | - | 8 - 9 | 1118 | 83 | ||
| AK145834.1 | - | 8 - 9 | 1109 | 83 | ||
| BC032994.1 | - | 1 - 2 | 58 | 83 | ||
| BC045199.1 | - | 8 - 9 | 1093 | 5133197 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acad11
Genomic Map