Assay Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009275.4 | NP_033301.1 | 4 - 5 | 415 | 68 | |
| GenBank mRNA | AK004854.1 | - | 4 - 5 | 448 | 68 | |
| AK007859.1 | - | 4 - 5 | 421 | 68 | ||
| AK008383.1 | - | 4 - 5 | 458 | 68 | ||
| AK015469.1 | - | 4 - 5 | 480 | 68 | ||
| AK030437.1 | - | 4 - 5 | 416 | 68 | ||
| AK075583.1 | - | 4 - 5 | 418 | 68 | ||
| AK160600.1 | - | 3 - 4 | 1649 | 68 | ||
| AK167039.1 | - | 4 - 5 | 421 | 68 | ||
| BC003798.1 | - | 4 - 5 | 436 | 3490766 | 68 | |
| U17343.1 | - | 4 - 5 | 429 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srprb
Genomic Map