Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 13 - 14 | 1693 | 90 | |
| GenBank mRNA | AF440692.1 | - | 12 - 13 | 1623 | 90 | |
| AK085754.1 | - | 13 - 14 | 1678 | 90 | ||
| AK142599.1 | - | 1683 | 90 | |||
| AK146549.1 | - | 13 - 14 | 1678 | 90 | ||
| AK149559.1 | - | 13 - 14 | 1678 | 90 | ||
| AK149595.1 | - | 13 - 14 | 1678 | 90 | ||
| AK168368.1 | - | 13 - 14 | 1680 | 90 | ||
| AK168405.1 | - | 13 - 14 | 1680 | 90 | ||
| AK168419.1 | - | 13 - 14 | 1678 | 90 | ||
| AK168938.1 | - | 13 - 14 | 1678 | 90 | ||
| AK169187.1 | - | 13 - 14 | 1677 | 90 | ||
| AK203903.1 | - | 1 - 2 | 100 | 90 | ||
| BC008559.1 | - | 11 - 12 | 1415 | 90 | ||
| BC012313.1 | - | 13 - 14 | 1693 | 3981648 | 90 | |
| BC018573.1 | - | 8 - 9 | 963 | 90 | ||
| BC020295.1 | - | 13 - 14 | 1662 | 90 | ||
| BC022986.1 | - | 13 - 14 | 1671 | 5123741 | 90 | |
| BC058216.1 | - | 13 - 14 | 1659 | 90 | ||
| BC058218.1 | - | 13 - 14 | 1672 | 90 | ||
| BC092046.1 | - | 13 - 14 | 1654 | 3594705 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trf
Genomic Map