Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 5 - 6 | 701 | 70 | |
| GenBank mRNA | AF440692.1 | - | 4 - 5 | 631 | 70 | |
| AK085754.1 | - | 5 - 6 | 686 | 70 | ||
| AK142599.1 | - | 691 | 70 | |||
| AK146549.1 | - | 5 - 6 | 686 | 70 | ||
| AK149559.1 | - | 5 - 6 | 686 | 70 | ||
| AK149595.1 | - | 5 - 6 | 686 | 70 | ||
| AK150782.1 | - | 5 - 6 | 734 | 70 | ||
| AK168368.1 | - | 5 - 6 | 687 | 70 | ||
| AK168405.1 | - | 5 - 6 | 687 | 70 | ||
| AK168419.1 | - | 5 - 6 | 686 | 70 | ||
| AK168938.1 | - | 5 - 6 | 686 | 70 | ||
| BC008559.1 | - | 3 - 4 | 423 | 70 | ||
| BC012313.1 | - | 5 - 6 | 701 | 3981648 | 70 | |
| BC020295.1 | - | 5 - 6 | 670 | 70 | ||
| BC022986.1 | - | 5 - 6 | 679 | 5123741 | 70 | |
| BC058218.1 | - | 5 - 6 | 680 | 70 | ||
| BC092046.1 | - | 5 - 6 | 662 | 3594705 | 70 | |
More Information
Set Membership: |
|

Trf
Genomic Map