Assay Details
| Gene Symbol: Mst1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287261.1 | NP_001274190.1 | 1 - 2 | 151 | 59 | |
| NM_009074.2 | NP_033100.2 | 11 - 12 | 3061 | 59 | ||
| NR_109782.1 | 11 - 12 | 3092 | 59 | |||
| GenBank mRNA | AK053655.1 | - | 11 - 12 | 3075 | 59 | |
| BC141379.1 | - | 11 - 12 | 2872 | 9055992 | 59 | |
| X74736.1 | - | 11 - 12 | 3061 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mst1r
Genomic Map