Assay Details
| Gene Symbol: Mst1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287261.1 | NP_001274190.1 | 3 - 4 | 546 | 99 | |
| NM_009074.2 | NP_033100.2 | 13 - 14 | 3456 | 99 | ||
| NR_109782.1 | 13 - 14 | 3487 | 99 | |||
| GenBank mRNA | AK053655.1 | - | 13 - 14 | 3470 | 99 | |
| BC141379.1 | - | 13 - 14 | 3267 | 9055992 | 99 | |
| X74736.1 | - | 13 - 14 | 3456 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mst1r
Genomic Map