Assay Details
| Gene Symbol: Fam212a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026597.3 | NP_080873.2 | 1 - 2 | 226 | 74 | |
| GenBank mRNA | AK020090.1 | - | 1 - 2 | 190 | 74 | |
| AK076092.1 | - | 1 - 2 | 190 | 74 | ||
| BC115538.1 | - | 1 - 2 | 165 | 40101503 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812297 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812301 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812300 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812299 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132772 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812304 | - | - |
| Whole Mouse Genome G4122A | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | BQ176112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | BC115538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | NM_026597.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | AK020090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248857 | AK076092.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07966650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08048264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08048266 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08048292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812300 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812301 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812297 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07966643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08048253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07753618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07636494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08048289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07840019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07966647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07831172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812304 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10596814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596813 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452903_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596812 | - | - |
| Mouse Genome 430 2.0 Array | 1452903_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452903_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68176_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68176_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900025588.mm.1 | NM_026597.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025588.mm.1 | NM_026597.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025588.mm.1 | TC0900003047.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025584.mm.1 | NM_026597.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025584.mm.1 | NM_026597.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025584.mm.1 | TC0900003047.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025586.mm.1 | NM_026597.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025586.mm.1 | NM_026597.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025586.mm.1 | TC0900003047.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025583.mm.1 | NM_026597.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025583.mm.1 | NM_026597.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025583.mm.1 | TC0900003047.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452903_at | - | - |
| Mouse Expression Set 430 | 1452903_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452903_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452903_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 371948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103940671 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661799 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2888375 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2888375 | - | - |

Fam212a
Genomic Map