Assay Details
| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133984.2 | NP_598745.2 | 3 - 4 | 541 | 72 | |
| GenBank mRNA | AK054445.1 | - | 3 - 4 | 492 | 72 | |
| AK088071.1 | - | 2 - 3 | 846 | 72 | ||
| BC011431.1 | - | 2 - 3 | 486 | 3981356 | 72 | |
| BC024428.1 | - | 2 - 3 | 346 | 4971671 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hemk1
Genomic Map