Assay Details
| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133984.2 | NP_598745.2 | 5 - 6 | 763 | 74 | |
| GenBank mRNA | AK088071.1 | - | 4 - 5 | 1068 | 74 | |
| BC011431.1 | - | 4 - 5 | 708 | 3981356 | 74 | |
| BC024428.1 | - | 4 - 5 | 568 | 4971671 | 74 | |
More Information
Set Membership: |
|

Hemk1
Genomic Map